5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)

Supplier: TCI America
Warning

B5036-1G B5036-5G
TCB5036-1GEA 83 USD
TCB5036-1G TCB5036-5G
5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)
5-Bromo-2-methylbenzaldehyde
Formula: C₈H₇BrO
Boiling Pt: 172°C
MDL Number: MFCD05662390
CAS Number: 90050-59-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 95.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR