2-Amino-4-phenylthiazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
A2846-1G
A2846-5G
TCA2846-1GEA
74.3
USD
TCA2846-1G
TCA2846-5G
2-Amino-4-phenylthiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-phenylthiazole
Formula:
C₉H₈N₂S |
MDL Number:
MFCD00039680 CAS Number: 2010-06-2 EINECS: 217-926-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...