1-Butyl-3-methylimidazolium hydrogen sulphate ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Danger
|
Synonyms:
3-Butyl-1-methyl-1H-imidazol-3-ium hydrogensulphate
B5569-25G
B5569-5G
TCB5569-25GEA
122
USD
TCB5569-25G
TCB5569-5G
1-Butyl-3-methylimidazolium hydrogen sulphate ≥97.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium hydrogen sulphate
Formula:
C₈H₁₆N₂O₄S MW: 236.29 Flash Pt: 284°C |
MDL Number:
MFCD06798197 CAS Number: 262297-13-2 UN: 3265 ADR: 8 II |
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...