2-Chloro-6-methylbenzothiazole ≥98.0% (by GC)

Supplier: TCI America

C3160-1G C3160-5G
TCC3160-1GEA 68 USD
TCC3160-1G TCC3160-5G
2-Chloro-6-methylbenzothiazole ≥98.0% (by GC)
2-Chloro-6-methylbenzothiazole
Formula: C₈H₆ClNS
Boiling Pt: 152°C
MDL Number: MFCD04448821
CAS Number: 3507-26-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR