1-Acetyl-5-bromoindoline ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(5-Bromoindolin-1-yl)ethanone

A2924-25G A2924-5G
76218-754EA 267 USD
76218-754 76218-756
1-Acetyl-5-bromoindoline ≥98.0% (by GC)
1-Acetyl-5-bromoindoline
Formula: C₁₀H₁₀BrNO
MW: 240.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00056017
CAS Number: 22190-38-1

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR