1-Hexyl-3-methylimidazolium iodide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 1-hexyl-3-methylimidazolium iodide

H1633-25G H1633-5G
76220-166EA 240 USD
76220-166 76220-168
1-Hexyl-3-methylimidazolium iodide ≥98.0% (by HPLC, total nitrogen)
1-Hexyl-3-methylimidazolium iodide
Formula: C₁₀H₁₉IN₂
MW: 294.18 g/mol
MDL Number: MFCD12964976
CAS Number: 178631-05-5

Order Now

Specification Test Results

Appearance White to Brown to Dark purple clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR