2-Bromo-4-chlorobenzaldehyde ≥97.0% (by GC)

Supplier: TCI America
Warning

B5524-25G B5524-5G
TCB5524-25GEA 182 USD
TCB5524-25G TCB5524-5G
2-Bromo-4-chlorobenzaldehyde ≥97.0% (by GC)
2-Bromo-4-chlorobenzaldehyde
Formula: C₇H₄BrClO
MDL Number: MFCD09056792
CAS Number: 84459-33-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR