2-Methylpyridin-3-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H1365-1G H1365-5G
TCH1365-1GEA 104 USD
TCH1365-1G TCH1365-5G
2-Methylpyridin-3-ol ≥98.0% (by GC, titration analysis)
2-Methylpyridin-3-ol
Formula: C₆H₇NO
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR