4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(Perfluorobutyl)-1-propanol, Daikin A-1430, 1H,1H,2H,2H,3H,3H-Nonafluoro-1-heptanol, 1H,1H,2H,2H,3H,3H-Perfluoro-1-heptanol, 4,4,5,5,6,6,7,7,7-Nonafluoroheptan-1-ol, 3-(Perfluorobutyl)propanol

N1040-100G N1040-25G N1040-5G
TCN1040-100GEA 933 USD
TCN1040-100G TCN1040-25G TCN1040-5G
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥98.0% (by GC)
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol
Formula: C₇H₇F₉O
MW: 278.12 g/mol
Boiling Pt: 64°C
Flash Pt: 76°C
MDL Number: MFCD02183552
CAS Number: 83310-97-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR