1,3-Butadiene ∼13% ∼2 mol/L in THF
Supplier: TCI America
|
Danger
|
Synonyms:
Buta-1,3-diene
B4835-100ML
B4835-500ML
TCB4835-100MLEA
78.5
USD
TCB4835-100ML
TCB4835-500ML
1,3-Butadiene ∼13% ∼2 mol/L in THF
1,3-Butadiene
Formula:
C₄H₆ MW: 54.09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00008659 CAS Number: 106-99-0 EINECS: 203-450-8 UN: 1993 ADR: 3,II Merck Index: 14,01509 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Concentration (HNMR) | 13.0 to 15.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...