1,3-Butadiene ∼15% in toluene

Supplier: TCI America
Danger

Synonyms: Buta-1,3-diene

B4359-100ML B4359-500ML
76219-388EA 83.95 USD
76219-388 76219-390
1,3-Butadiene ∼15% in toluene
1,3-Butadiene
Formula: C₄H₆
MW: 54.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008659
CAS Number: 106-99-0
EINECS: 203-450-8
UN: 1993
ADR: 3,II
Merck Index: 14,01509

Order Now

Specification Test Results

Concentration (HNMR) 15.0 to 20.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR