1,3-Butadiene ∼13% in hexane

Supplier: TCI America
Danger

Synonyms: Buta-1,3-diene

B4358-100ML B4358-500ML
TCB4358-100MLEA 99.39 USD
TCB4358-100ML TCB4358-500ML
1,3-Butadiene ∼13% in hexane
1,3-Butadiene
Formula: C₄H₆
MW: 54.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008659
CAS Number: 106-99-0
EINECS: 203-450-8
UN: 1993
ADR: 3,II
Merck Index: 14,01509

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 15.0 to 18.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR