2-Hydroxy-3-nitro-6-picoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Methyl-3-nitro-2(1H)-pyridinone, 2-Hydroxy-6-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-2-picoline, 6-Methyl-3-nitropyridin-2-ol, 6-Methyl-3-nitro-2-pyridone, 2-Methyl-5-nitro-6-pyridone, 6-Hydroxy-2-methyl-5-nitropyridine

H1498-1G H1498-5G
TCH1498-1GEA 19 USD
TCH1498-1G TCH1498-5G
2-Hydroxy-3-nitro-6-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-nitro-6-picoline
Formula: C₆H₆N₂O₃
MW: 154.13
Melting Pt: 217°C
MDL Number: MFCD03001414
CAS Number: 39745-39-6

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR