3,4-Difluorobenzenesulfonamide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4892-1G
D4892-5G
TCD4892-1GEA
31.4
USD
TCD4892-1G
TCD4892-5G
3,4-Difluorobenzenesulfonamide ≥98.0% (by GC)
3,4-Difluorobenzenesulphonamide
Formula:
C₆H₅F₂NO₂S |
MDL Number:
MFCD00728800 CAS Number: 108966-71-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...