4-Fluoro-3-nitrobenzaldehyde ≥95.0% (by GC)

Supplier: TCI America
Warning

F0956-1G F0956-5G
TCF0956-1GEA 24 USD
TCF0956-1G TCF0956-5G
4-Fluoro-3-nitrobenzaldehyde ≥95.0% (by GC)
4-Fluoro-3-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.11 g/mol
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR