N,N-Diethylbenzamide ≥95.0% (by GC)

Supplier: TCI America
Warning

D5123-100G D5123-25G
TCD5123-100GEA 172 USD
TCD5123-100G TCD5123-25G
N,N-Diethylbenzamide ≥95.0% (by GC)
N,N-Diethylbenzamide
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 280°C
MDL Number: MFCD00026726
CAS Number: 1696-17-9
EINECS: 216-912-9

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR