4',6,7-Trimethoxyisoflavone ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
6,7-Dimethoxy-3-(4-methoxyphenyl)chromone
T3410-1G
T3410-5G
TCT3410-1GEA
174
USD
TCT3410-1G
TCT3410-5G
4',6,7-Trimethoxyisoflavone ≥97.0% (by GC)
4',6,7-Trimethoxyisoflavone
Formula:
C₁₈H₁₆O₅ MW: 312.32 g/mol |
MDL Number:
MFCD00016949 CAS Number: 798-61-8 |
Specification Test Results
Purity (GC) | min. 97.0 % |
Melting point | 177.0 to 181.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...