Acacetin ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A3013-100MG
A3013-25MG
TCA3013-100MGEA
328
USD
TCA3013-100MG
TCA3013-25MG
Acacetin ≥98.0% (by GC)
Acacetin
Formula:
C₁₆H₁₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00016936 CAS Number: 480-44-4 EINECS: 207-552-3 Merck Index: 14,00013 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...