5-Chloro-2-methoxybenzenesulphonyl chloride ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

C2740-25G C2740-5G
TCC2740-25GEA 304 USD
TCC2740-25G TCC2740-5G
5-Chloro-2-methoxybenzenesulphonyl chloride ≥95.0% (by GC, titration analysis)
5-Chloro-2-methoxybenzenesulphonyl chloride
Formula: C₇H₆Cl₂O₃S
MDL Number: MFCD00052959
CAS Number: 22952-32-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR