2-Chloro-5-(trifluoromethyl)phenol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C2947-25G
C2947-5G
TCC2947-25GEA
154.8
USD
TCC2947-25G
TCC2947-5G
2-Chloro-5-(trifluoromethyl)phenol ≥98.0% (by GC)
2-Chloro-5-(trifluoromethyl)phenol
Formula:
C₇H₄ClF₃O Boiling Pt: 88°C Flash Pt: 46°C |
MDL Number:
MFCD00019995 CAS Number: 40889-91-6 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...