3-Chloro-5-fluoroaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
C3052-1G
C3052-5G
TCC3052-1GEA
55
USD
TCC3052-1G
TCC3052-5G
3-Chloro-5-fluoroaniline ≥97.0% (by GC, titration analysis)
3-Chloro-5-fluoroaniline
Formula:
C₆H₅ClFN Boiling Pt: 98°C |
MDL Number:
MFCD03407962 CAS Number: 4863-91-6 |
Specification Test Results
Appearance | Orange to Amber to Dark red clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...