Methyl-2-chloro-4-fluorobenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M3102-25G M3102-5G
TCM3102-25GEA 114 USD
TCM3102-25G TCM3102-5G
Methyl-2-chloro-4-fluorobenzoate ≥98.0% (by GC)
Methyl-2-chloro-4-fluorobenzoate
Formula: C₈H₆ClFO₂
MDL Number: MFCD00173937
CAS Number: 85953-29-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR