2-Chloro-5-fluoro-3-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-5-fluoro-3-picoline

C3144-1G C3144-5G
TCC3144-1GEA 42.9 USD
TCC3144-1G TCC3144-5G
2-Chloro-5-fluoro-3-methylpyridine ≥98.0% (by GC)
2-Chloro-5-fluoro-3-methylpyridine
Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD04972725
CAS Number: 38186-84-4

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR