6-Chloroflavone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Chloro-2-phenyl-4H-benzo[b]pyran-4-one, 6-Chloro-2-phenylchromen-4-one, 6-Chloro-2-phenyl-4H-1-benzopyran-4-one, 6-Chloro-2-phenyl-4H-chromen-4-one, 6-Chloro-2-phenyl-4-benzopyrone

C3066-1G C3066-5G
TCC3066-1GEA 106 USD
TCC3066-1G TCC3066-5G
6-Chloroflavone ≥98.0% (by GC)
6-Chloroflavone
Formula: C₁₅H₉ClO₂
MW: 256.69 g/mol
MDL Number: MFCD00209560
CAS Number: 10420-73-2

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR