3-Chloro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3083-1G C3083-5G
TCC3083-1GEA 37 USD
TCC3083-1G TCC3083-5G
3-Chloro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
3-Chloro-4-hydroxybenzonitrile
Formula: C₇H₄ClNO
MDL Number: MFCD01567246
CAS Number: 2315-81-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR