1-Bromo-3-fluoro-2-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-6-fluoronitrobenzene

B4842-1G B4842-5G
TCB4842-1GEA 21 USD
TCB4842-1G TCB4842-5G
1-Bromo-3-fluoro-2-nitrobenzene ≥98.0% (by GC)
1-Bromo-3-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368788
CAS Number: 886762-70-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR