1-Bromo-2,4-difluoro-5-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2,4-difluoronitrobenzene

B5688-1G B5688-5G
76218-884EA 21 USD
76218-884 76218-886
1-Bromo-2,4-difluoro-5-nitrobenzene ≥98.0% (by GC)
1-Bromo-2,4-difluoro-5-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
Boiling Pt: 97°C
Melting Pt: 19°C
MDL Number: MFCD00129166
CAS Number: 345-24-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR