2-Bromo-5-chloro-3-methylpyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5133-25G
B5133-5G
TCB5133-25GEA
197
USD
TCB5133-25G
TCB5133-5G
2-Bromo-5-chloro-3-methylpyridine ≥98.0% (by GC)
2-Bromo-5-chloro-3-methylpyridine
Formula:
C₆H₅BrClN |
MDL Number:
MFCD03095062 CAS Number: 65550-77-8 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...