Resazurin sodium salt ≥85.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

R0203-1G R0203-5G
TCR0203-1GEA 34.58 USD
TCR0203-1G TCR0203-5G
Resazurin sodium salt ≥85.0% (by HPLC)
Resazurin sodium salt
Formula: C₁₂H₆NNaO₄
MW: 251.17 g/mol
MDL Number: MFCD00005036
CAS Number: 62758-13-8
EINECS: 263-718-5
Merck Index: 14,08141

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 85.0 area%
Lambda max. 598.0 to 602.0nm (H2O)
Absorbance min. 0.450 (5mg/L、H2O、598.0 to 602.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR