4'-Chloro-2'-hydroxyacetophenone ≥96.0% (by GC)

Supplier: TCI America
Warning

C2968-25G C2968-5G
TCC2968-25GEA 175.8 USD
TCC2968-25G TCC2968-5G
4'-Chloro-2'-hydroxyacetophenone ≥96.0% (by GC)
4'-Chloro-2'-hydroxyacetophenone
Formula: C₈H₇ClO₂
Boiling Pt: 122°C
MDL Number: MFCD00238557
CAS Number: 6921-66-0

Order Now

Specification Test Results

Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR