4-Fluoro-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

F0979-25G F0979-5G
TCF0979-25GEA 195.6 USD
TCF0979-25G TCF0979-5G
4-Fluoro-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MDL Number: MFCD00061284
CAS Number: 67515-59-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR