Methyl-3-nitrosalicylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 2-hydroxy-3-nitrobenzoate

M2864-1G M2864-5G
TCM2864-1GEA 27 USD
TCM2864-1G TCM2864-5G
Methyl-3-nitrosalicylate ≥98.0% (by GC, titration analysis)
Methyl-3-nitrosalicylate
Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD00272263
CAS Number: 22621-41-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR