4-Nitro-o-toluonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

M2965-1G M2965-5G
TCM2965-1GEA 98 USD
TCM2965-1G TCM2965-5G
4-Nitro-o-toluonitrile ≥98.0% (by GC)
4-Nitro-o-toluonitrile
Formula: C₈H₆N₂O₂
MDL Number: MFCD03095378
CAS Number: 89001-53-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR