2-Amino-3,5-dichlorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

A3062-1G A3062-5G
TCA3062-1GEA 87 USD
TCA3062-1G TCA3062-5G
2-Amino-3,5-dichlorobenzonitrile ≥98.0% (by GC)
2-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00158947
CAS Number: 36764-94-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR