4-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Chloro-2-fluorobenzeneboronic acid, (4-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(4-chloro-2-fluorophenyl)borane

C3048-1G C3048-5G
TCC3048-1GEA 31.4 USD
TCC3048-1G TCC3048-5G
4-Chloro-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MW: 174.37
Melting Pt: 250°C
MDL Number: MFCD02684293
CAS Number: 160591-91-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR