4-Bromo-2,6-difluorobenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
(4-Bromo-2,6-difluorophenyl)methanol
B5090-1G
B5090-5G
TCB5090-1GEA
53
USD
TCB5090-1G
TCB5090-5G
4-Bromo-2,6-difluorobenzyl alcohol ≥98.0% (by GC)
4-Bromo-2,6-difluorobenzyl alcohol
Formula:
C₇H₅BrF₂O MW: 223.02 g/mol |
MDL Number:
MFCD03094461 CAS Number: 162744-59-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...