4-Amino-2-chloropyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Chlor-4-pyrimidinamin, 2-Chloro-4-pyrimidinamine, 2-Chloropyrimidin-4-amine, 2-Chloropyrimidin-4-ylamine

A2719-1G A2719-5G
TCA2719-1GEA 40 USD
TCA2719-1G TCA2719-5G
4-Amino-2-chloropyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2-chloropyrimidine
Formula: C₄H₄ClN₃
MW: 129.55
MDL Number: MFCD00194051
CAS Number: 7461-50-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR