4-Isopropoxybenzaldehyde ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: ALDH1A3-IN-3

I0949-25G I0949-5G
TCI0949-25GEA 168 USD
TCI0949-25G TCI0949-5G
4-Isopropoxybenzaldehyde ≥96.0% (by GC)
4-Isopropoxybenzaldehyde
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 112°C
MDL Number: MFCD00052357
CAS Number: 18962-05-5

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR