3-Amino-4-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2971-1G A2971-5G
TCA2971-1GEA 124 USD
TCA2971-1G TCA2971-5G
3-Amino-4-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
3-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MDL Number: MFCD00173960
CAS Number: 175204-80-5

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR