3-(2-Methoxyphenyl)propionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2673-25G M2673-5G
TCM2673-25GEA 294 USD
TCM2673-25G TCM2673-5G
3-(2-Methoxyphenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(2-Methoxyphenyl)propionic acid
Formula: C₁₀H₁₂O₃
MDL Number: MFCD00002772
CAS Number: 6342-77-4
EINECS: 228-738-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR