2-Phenylbenzoxazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Phenyl-1,3-benzoxazole, 2-Phenylbenzo[d]oxazole

P2264-1G P2264-5G
TCP2264-1GEA 102 USD
TCP2264-1G TCP2264-5G
2-Phenylbenzoxazole ≥98.0% (by GC)
2-Phenylbenzoxazole
Formula: C₁₃H₉NO
MW: 195.22 g/mol
MDL Number: MFCD00012183
CAS Number: 833-50-1

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR