2-Fluoro-6-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

F1033-1G F1033-5G
TCF1033-1GEA 47.1 USD
TCF1033-1G TCF1033-5G
2-Fluoro-6-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
2-Fluoro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 67°C
Flash Pt: 82°C
MDL Number: MFCD00061273
CAS Number: 60611-24-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR