2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)

Supplier: TCI America
Warning

B4508-1G B4508-5G
TCB4508-1GEA 31 USD
TCB4508-1G TCB4508-5G
2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
2-Bromo-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD04973760
CAS Number: 102684-91-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR