4,4'-Dimethylbenzhydrol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Bis(4-methylphenyl)methanol, 4,4'-Dimethylbenzhydryl alcohol, Di-p-tolylmethanol, 4,4'-Dimethyl-α-phenylbenzyl alcohol, Bis(4-methylphenyl)carbinol, 4,4'-Dimethylphenyl phenyl carbinol, 4,4'-Dimethyldiphenylmethanol

D5460-1G D5460-5G
TCD5460-1GEA 85 USD
TCD5460-1G TCD5460-5G
4,4'-Dimethylbenzhydrol ≥98.0% (by GC)
4,4'-Dimethylbenzhydrol
Formula: C₁₅H₁₆O
MW: 212.29 g/mol
MDL Number: MFCD00017216
CAS Number: 885-77-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR