1-Benzyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5726-25G B5726-5G
TCB5726-25GEA 214 USD
TCB5726-25G TCB5726-5G
1-Benzyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
1-Benzyl-3-methylimidazolium hexafluorophosphate
Formula: C₁₁H₁₃F₆N₂P
MW: 318.2
MDL Number: MFCD07784448
CAS Number: 433337-11-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR