1-Ethyl-3-methylimidazolium nitrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

E0775-25G E0775-5G
TCE0775-25GEA 240 USD
TCE0775-25G TCE0775-5G
1-Ethyl-3-methylimidazolium nitrate ≥98.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium nitrate
Formula: C₆H₁₁N₃O₃
MW: 173.17 g/mol
Melting Pt: 41°C
MDL Number: MFCD00210007
CAS Number: 143314-14-1
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR