1-Decyl-3-methylimidazolium bromide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-Decyl-3-methyl-1H-imidazol-3-ium bromide

D5350-25G D5350-5G
76219-936EA 275 USD
76219-936 76219-938
1-Decyl-3-methylimidazolium bromide ≥98.0% (by HPLC)
1-Decyl-3-methylimidazolium bromide
Formula: C₁₄H₂₇BrN₂
MW: 303.29 g/mol
MDL Number: MFCD09038832
CAS Number: 188589-32-4

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR