1-Butyl-3-methylimidazolium methyl sulphate ≥98.0% (by HPLC)
Supplier: TCI America
|
Danger
|
Synonyms:
[BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4
B5527-25G
B5527-5G
TCB5527-25GEA
174
USD
TCB5527-25G
TCB5527-5G
1-Butyl-3-methylimidazolium methyl sulphate ≥98.0% (by HPLC)
1-Butyl-3-methylimidazolium methyl sulphate
Formula:
C₉H₁₈N₂O₄S Melting Pt: -4°C |
MDL Number:
MFCD03095437 CAS Number: 401788-98-5 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...