4-Bromobenzhydrol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: p-Bromobenzhydrol, 4-Bromophenyl phenyl carbinol, 4-Bromodiphenylmethanol, 4-Bromo-α-phenylbenzyl alcohol, 4-Bromobenzhydryl alcohol, (4-Bromophenyl)(phenyl)methanol

B4680-1G B4680-5G
TCB4680-1GEA 46 USD
TCB4680-1G TCB4680-5G
4-Bromobenzhydrol ≥97.0% (by GC)
4-Bromobenzhydrol
Formula: C₁₃H₁₁BrO
MW: 263.13
MDL Number: MFCD00016851
CAS Number: 29334-16-5
EINECS: 249-568-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR