6-Bromo-m-toluonitrile ≥97.0% (by GC)

Supplier: TCI America
Danger

B4678-1G B4678-5G
TCB4678-1GEA 43 USD
TCB4678-1G TCB4678-5G
6-Bromo-m-toluonitrile ≥97.0% (by GC)
6-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MDL Number: MFCD11040281
CAS Number: 42872-83-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR